Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium Wire, 0.25mm (0.01 in.) dia., 99.9% (Metals basis), Alfa Aesar™

Click to view available options
Quantity:
1m
5m
7440-05-3
Pd
106.42
MFCD00011167
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
7440-05-3
Pd
106.42
MFCD00011167
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]

Form: Approx.0.6g/m

Quantity 1m
Appearance Silver gray wire
Assay Percent Range 99.9% (Metals basis)
Odor Odorless
Physical Form Wire, ≈0.6g/m
Color Silver to Gray
Chemical Name or Material Palladium

EINECSNumber : 231-115-6

TSCA : Yes

Recommended Storage : Ambient temperatures