Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Pallbearer Gloves
SDP

Catalog No. 22445445 Shop All Mopec Products
Click to view available options
:
Pallbearer Gloves; Color: White

Pallbearer Gloves made of cotton.

  • One size fits most
  • White color

Specifications

Applications Industrial
Coating Material Uncoated
Color White
Disposable Single-use
Exterior Finish Smooth
Hand Ambidextrous
Laboratory/Cleanroom Use No
Latex Free Yes
Material Cotton
Powdered Powder Free
Product Type Pallbearer Gloves
Size Universal
Sterility Non-sterile
Finger Style Full
Glove Cuff Style Hemmed
Quantity 12 Gloves/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.