Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PALLD/palladin Mouse anti-Human, Porcine, Rat, Clone: 2A1F6, Proteintech
SDP

Catalog No. 501737019 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7019 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7019 Supplier Proteintech Group Inc Supplier No. 666011IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Palladin is a cytoskeletal protein required for organization of normal actin cytoskeleton. Palladin plays roles in establishing cell morphology, motility, cell adhesion and cell-extracellular matrix interactions in a variety of cell types. Palladin may function as a scaffolding molecule with the potential to impact both actin polymerization as well as the assembly of existing actin filaments into higher-order arrays. It binds to proteins that bind to either monomeric or filamentous actin. It localizes at sites where active actin remodeling takes place, such as lamellipodia and membrane ruffles. Palladin′s different isoforms may have functional differences. It is also involved in the control of morphological and cytoskeletal changes associated with dendritic cell maturation.

Specifications

Antigen PALLD/palladin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2A1F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PALLD
Gene Accession No. P0C5E3, Q8WX93
Gene Alias CGI 151, KIAA0992, palladin, PALLD, PNCA1, Sarcoma antigen NY SAR 77, SIH002
Gene Symbols Palld
Host Species Mouse
Immunogen PALLD,palladin Fusion Protein Ag1288
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100627292, 23022, 364558
Target Species Human, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.