Learn More
Invitrogen™ Pallidin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114040
Description
Immunogen sequence: EGLIEDLTIE DKAVEQLAEG LLSHYLPDLQ RSKQALQELT QNQVVLLDTL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The aquaporins are a family of water-selective membrane channels. This gene encodes a member of a subset of aquaporins called the aquaglyceroporins. This protein allows passage of a broad range of noncharged solutes and also stimulates urea transport and osmotic water permeability. This protein may also facilitate the uptake of glycerol in hepatic tissue. The encoded protein may also play a role in specialized leukocyte functions such as immunological response and bactericidal activity. Alternate splicing results in multiple transcript variants.
Specifications
| Pallidin | |
| Polyclonal | |
| Unconjugated | |
| BLOC1S6 | |
| biogenesis of lysosomal organelles complex 1 subunit 6; biogenesis of lysosomal organelles complex-1, subunit 5, pallidin; biogenesis of lysosomal organelles complex-1, subunit 6, pallidin; biogenesis of lysosome-related organelles complex 1 subunit 6; biogenesis of organelles complex-1, subunit 6, pallidin; BLOC-1; BLOC-1 subunit 6; Bloc1s6; BLOS6; HPS9; P2; PA; PALLID; Pallid protein; pallid protein homolog; Pallidin; pallidin homolog; PLDN; Stx13bp1; syntaxin 13 binding protein 1; Syntaxin 13-interacting protein; syntaxin 13-interacting protein pallid | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26258 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UL45 | |
| BLOC1S6 | |
| Synthetic peptide directed towards the middle region of human PLDN. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.