Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PALM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559690
Description
Immunogen sequence: TKKVLGLQDT ITAELVVIED AAEPKEPAPP NGSAAEPPTE AASREENQAG PEATTSDPQD LDMKKHRCKC CSIM Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 73%.
This gene encodes a member of the paralemmin protein family. The product of this gene is a prenylated and palmitoylated phosphoprotein that associates with the cytoplasmic face of plasma membranes and is implicated in plasma membrane dynamics in neurons and other cell types. Several alternatively spliced transcript variants have been identified, but the full-length nature of only two transcript variants has been determined.
Specifications
PALM | |
Polyclonal | |
Unconjugated | |
PALM | |
KIAA0270; Palm; Paralemmin; paralemmin-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5064 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75781 | |
PALM | |
Recombinant protein corresponding to Human PALM. Recombinant protein control fragment (Product #RP-98912). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction