Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PALM2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567333
Description
Immunogen sequence: TKVVYEVRSGG TVVENGVHKL STKDVEELIQ KAGQSSLGGG HVSERTVIAD GSLSHPKEHM LCKEAKLEMV HKSRKDHSSG NPGQQ Highest antigen sequence identity to the following orthologs - mouse 87%, rat 87%.
PALM2 gene ontology annotations related to this gene include regulation of cell shape.
Specifications
PALM2 | |
Polyclonal | |
Unconjugated | |
PALM2 | |
A kinase (PRKA) anchor protein 2; AKAP2; PALM2; paralemmin 2; paralemmin-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114299 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IXS6 | |
PALM2 | |
Recombinant Human PALM2. Recombinant protein control fragment (Product #RP-105666). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction