Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PALM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572694
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572694 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572694 Supplier Invitrogen™ Supplier No. PA572694
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PALM3 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

ATP-binding protein, which may act as a adapter in the Toll-like receptor (TLR) signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PALM3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PALM3
Gene Accession No. A6NDB9
Gene Alias PALM3; paralemmin 3; paralemmin-3
Gene Symbols PALM3
Host Species Rabbit
Immunogen Paralemmin3 antibody for a 19 amino acid synthetic peptide near the amino terminus of human Paralemmin3. The immunogen is within amino acids 50 - 100 of Paralemmin3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342979
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.