Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PALMD Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PALMD |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PALMD Polyclonal specifically detects PALMD in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PALMD | |
Polyclonal | |
Rabbit | |
Human | |
54873 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SGKEQEEMKKQNQQDQHQIQVLEQSILRLEKEIQDLEKAELQISTKEEAILKKLKSIERTTEDIIRSVKVEREERAEESIEDIYANIPDLPKSYIPSRLRKEINEEKEDDEQNRKALYAMEIKVEKDLKTGEST | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
C1orf11, palmdelphin, PALMLFLJ20271, Paralemnin-like protein | |
PALMD | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title