Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PALMD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565070
Description
Immunogen sequence: SGKEQEEMKKQ NQQDQHQIQV LEQSILRLEK EIQDLEKAEL QISTKEEAIL KKLKSIERTT EDIIRSVKVE REERAEESIE DIYANIPDLP KSYIPSRLRK EINEEKEDDE QNRKALYAME IKVEKDLKTG EST Highest antigen sequence identity to the following orthologs - mouse 90%, rat 54%.
PALMD gene ontology annotations related to this gene include regulation of cell shape.
Specifications
PALMD | |
Polyclonal | |
Unconjugated | |
PALMD | |
4631423C22Rik; C1orf11; PALMD; Palmdelphin; PALML; Paralemmin-like protein; paralemmin-related; paralemnin-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54873 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NP74 | |
PALMD | |
Recombinant Human PALMD. Recombinant protein control fragment (Product #RP-105832). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction