Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PALMD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572871
Description
PALMD Polyclonal Antibody for Western Blot, IHC (P), ELISA
PALMD gene ontology annotations related to this gene include regulation of cell shape.
Specifications
PALMD | |
Polyclonal | |
Unconjugated | |
PALMD | |
4631423C22Rik; C1orf11; PALMD; Palmdelphin; PALML; Paralemmin-like protein; paralemmin-related; paralemnin-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54873 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9NP74 | |
PALMD | |
PALMD antibody for a 15 amino acid peptide near the carboxy terminus of human PALMD. The immunogen is within amino acids 460 - 510 of PALMD. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction