Learn More
Palmitic Acid, [1-14C], MP Biomedicals™
Catalog No. MP011219583 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 mci; 37 mbq
250 μci; 9.25 mbq
50 μci; 1.85 mbq
Molecular Formula CH3(CH2)14COOH
  • Sp. Act. 40-60 mCi/mmol; 1.48-2.22 GBq/mmol
  • Ethanol
TRUSTED_SUSTAINABILITY
Chemical Identifiers
Molecular Formula CH3(CH2)14COOH
Specifications
Quantity 250 μci; 9.25 mbq
storageNote1 Store at 2-8°C.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.