Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(+)-Tartaric acid, disodium salt, dihydrate, 99+%, ACS reagent

Catalog No. AC424440025
Click to view available options
Quantity:
100 g
2.5 kg
10 kg

CAS: 6106-24-7 | C4H8Na2O8 | 230.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-24-7
Molecular Formula C4H8Na2O8
Molecular Weight (g/mol) 230.08
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
Synonym unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
PubChem CID 131855972
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

pH 7.0 to 9.0 (5% soln. at 25°C)
Color White
Assay Percent Range 99+%
Linear Formula (CHOHCOONa)2·2H2O
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: insolubel in organic solvents
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
Formula Weight 230.08
Percent Purity ≥99%
Grade ACS Reagent
Quantity 2.5 kg
Infrared Spectrum Authentic
Loss on Drying 15.61% to 15.71% (150°C)
Packaging Plastic bottle
Physical Form Needles or Powder
Chemical Name or Material L(+)-Tartaric acid, disodium salt, dihydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.