Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palmitoleic Acid Methyl Ester, MP Biomedicals™

Catalog No. mp215604890
Click to view available options
Quantity:
500 mg
1 g

Palmitoleic Acid Methyl Ester, MP Biomedicals is a component in the preparation of biodiesel fuels.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-25-8
Molecular Formula C17H32O2
Molecular Weight (g/mol) 268.44
MDL Number MFCD00042911
InChI Key IZFGRAGOVZCUFB-HJWRWDBZSA-N
Synonym methyl palmitoleate, z-methyl hexadec-9-enoate, methyl z-hexadec-9-enoate, methyl palmitoleinate, unii-35695qdb9f, methyl 9z-hexadec-9-enoate, 9-hexadecenoic acid, methyl ester, z, methyl cis-9-hexadecenoate, methyl z-9-hexadecenoate, methyl cis-hexadec-9-enoate
PubChem CID 643801
ChEBI CHEBI:84156
IUPAC Name methyl (9Z)-hexadec-9-enoate
SMILES CCCCCC\C=C/CCCCCCCC(=O)OC

Specifications

Melting Point -0.5°C to +0.5°C (literature)
Density 0.875g/mL at 25°C (Lit.)
Boiling Point 180°C to 183°C (1 mmHg)(literature)
Refractive Index n20/D 1.451 (Lit.)
Quantity 500 mg
Formula Weight 268.44
Physical Form Liquid
Recommended Storage −20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.