Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palmitoyl-L-carnitine-(N-methyl-d3), ≥97.0% (TLC), MilliporeSigma™ Supelco™

Catalog No. p-7187751 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
5 mg

Specifications

CAS 202480-73-7
Linear Formula C23D3H42 NO4
MDL Number MFCD01073471
Synonym (2 R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxohexadecyl)oxy]-1-propanaminium inner salt; L-Carnitine-(methyl-d3) hexadecanoyl ester; Hexadecanoyl-L-carnitine-(N-methyl-d3)
Type Analytical Standard
Quantity 5 mg
Formula Weight 402.63
Percent Purity ≥97.0% (TLC)
Grade Analytical Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.