Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PALS1/MPP5 Antibody (OTI2D3), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273234MFV610
Description
PALS1/MPP5 Monoclonal antibody specifically detects PALS1/MPP5 in Human, Mouse, Rat, Canine samples. It is validated for Flow CytometrySpecifications
PALS1/MPP5 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
64398 | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
OTI2D3 | |
Flow Cytometry | |
MAGUK p55 subfamily member 5, membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5), PALS1FLJ12615, stardust | |
Full length human recombinant protein of human MPP5(NP_071919) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction