Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEGAComfort PAM™ ESD Anti-Static Insole

Personal anti-fatigue mat ESD insole ergonomically designed by podiatrists, Dual layer 100% memory foam, Clinically proven field tested to reduce pain and fatigue, Max shock absorption

$29.95

Specifications

Color Black, Blue, Yellow
Material Memory Foam, ESD Thread
Product Type Specs ESD insole
Waterproof No
For Use With (Application) Stationary and mobile workers
View More Specs

Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
18-813-499
View Documents
MEGAComfort
PAMESDW567
Women's: 5-7
Pair for $29.95
Add to cart
 
18-813-500
View Documents
MEGAComfort
PAMESDM67/W89
Men's: 6-7, Women's: 8-9
Pair for $29.95
Add to cart
 
18-813-501
View Documents
MEGAComfort
PAMESDM89/W1011
Men's: 8-9, Women's: 10-11
Pair for $29.95
Add to cart
 
18-813-502
View Documents
MEGAComfort
PAMESDM1011/W1213
Men's: 10-11, Women's: 12-13
Pair for $29.95
Add to cart
 
18-813-503
View Documents
MEGAComfort
PAMESDM1213
Men's: 12-13
Pair for $29.95
Add to cart
 
18-813-504
View Documents
MEGAComfort
PAMESDM1415
Men's: 14-15
Pair for $29.95
Add to cart
 
Description

Description

  • Personal Anti-Fatigue Mat (PAM™) ESD insole
  • Uses patented combination of high density dual layer 100% memory foam
  • For anti-static footwear only
  • ESD threads provide tested electro-static dissipation
  • Reduces pain and fatigue while increasing balance and comfort
  • Ideal for stationary and mobile workers
  • ESD anti-static threads provide electro-static dissipation
  • Silver ion anti-microbial technology in top cloth offering anti-bacterial, anti-perspiration and anti-odor
  • Machine washable

Note:

  • PAM™ ESD does not make regular footwear ESD compliant
Specifications

Specifications

Black, Blue, Yellow
ESD insole
Stationary and mobile workers
Memory Foam, ESD Thread
No
Boots
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit