Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115606
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115606 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115606 Supplier Invitrogen™ Supplier No. PA5115606
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MYCBP2.

This gene encodes an E3 ubiquitin-protein ligase and member of the PHR (Phr1/MYCBP2, highwire and RPM-1) family of proteins. The encoded protein plays a role in axon guidance and synapse formation in the developing nervous system. In mammalian cells, this protein regulates the cAMP and mTOR signaling pathways, and may additionally regulate autophagy. Reduced expression of this gene has been observed in acute lymphoblastic leukemia patients and a mutation in this gene has been identified in patients with a rare inherited vision defect.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAM
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene MYCBP2
Gene Accession No. O75592, Q7TPH6
Gene Alias AU023734; AW107953; AW546647; C130061D10Rik; E3 ubiquitin-protein ligase MYCBP2; Highwire; KIAA0916; MYC binding protein 2; MYC binding protein 2, E3 ubiquitin protein ligase; Myc-binding protein 2; Mycbp2; Myc-bp2; PAM; pam, highwire, rpm 1; pam/highwire/rpm-1 protein; Phr; Phr1; probable E3 ubiquitin-protein ligase MYCBP2; protein associated with Myc; Protein Magellan; R75243
Gene Symbols MYCBP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human MYCBP2(Accession O75592), corresponding to amino acid residues S979-E1029.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105689, 23077, 290447
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.