Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAM16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597278 Supplier Invitrogen™ Supplier No. PA597278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARALRQEFAA SRAAADARGR AGHRSAAASN LSGLSLQEAQ QILNVSKLSP EEVQKNYEHL FKVNDKSVGG SFYLQSKVVR AKERLDEELK IQAQEDREKG QMPHT; Positive Samples: Jurkat, Mouse testis; Cellular Location: Matrix side, Mitochondrion inner membrane, Peripheral membrane protein.

Regulates ATP-dependent protein translocation into the mitochondrial matrix. Inhibits DNAJC19 stimulation of HSPA9/Mortalin ATPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAM16
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PAM16
Gene Accession No. Q9CQV1, Q9Y3D7
Gene Alias 2010110I09Rik; AV006767; CGI-136; MAGMAS; magmas processed gene 1; magmas-like protein; mitochondria associated granulocyte macrophage CSF signaling molecule; mitochondria associated protein involved in granulocyte macrophage colony stimulating factor signal transduction; mitochondria-associated granulocyte macrophage CSF-signaling molecule; mitochondria-associated protein involved in granulocyte-macrophage colony-stimulating factor signal transduction; mitochondrial import inner membrane translocase subunit TIM16; Pam16; presequence translocase-associated motor 16 homolog (S. cerevisiae); presequence translocase-asssociated motor 16 homolog (S. cerevisiae); presequence translocated-associated motor subunit PAM16; SMDMDM; TIM16; Timm16
Gene Symbols PAM16
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 21-125 of human PAM16 (NP_0571538).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51025, 66449
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.