Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAM16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599082
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599082 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599082 Supplier Invitrogen™ Supplier No. PA599082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PAM16 Polyclonal Antibody for Western Blot, ELISA

Regulates ATP-dependent protein translocation into the mitochondrial matrix. Inhibits DNAJC19 stimulation of HSPA9/Mortalin ATPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAM16
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PAM16
Gene Accession No. Q9Y3D7
Gene Alias 2010110I09Rik; AV006767; CGI-136; MAGMAS; magmas processed gene 1; magmas-like protein; mitochondria associated granulocyte macrophage CSF signaling molecule; mitochondria associated protein involved in granulocyte macrophage colony stimulating factor signal transduction; mitochondria-associated granulocyte macrophage CSF-signaling molecule; mitochondria-associated protein involved in granulocyte-macrophage colony-stimulating factor signal transduction; mitochondrial import inner membrane translocase subunit TIM16; Pam16; presequence translocase-associated motor 16 homolog (S. cerevisiae); presequence translocase-asssociated motor 16 homolog (S. cerevisiae); presequence translocated-associated motor subunit PAM16; SMDMDM; TIM16; Timm16
Gene Symbols PAM16
Host Species Rabbit
Immunogen Recombinant Human Mitochondrial import inner membrane translocase subunit TIM16 protein (1-125AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51025
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.