Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAMR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PAMR1 |
---|---|
Concentration | 0.24 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6106
![]() ![]() |
Proteintech Group Inc
553101AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is specific to PAMR1.
PAMR1 may be involved in regeneration of skeletal muscle. Although related to peptidase S1 family, lacks the conserved active Ser residue in position 665 which is replaced by a Thr, suggesting that it has no protease activity.Specifications
PAMR1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q6UXH9, Q8BU25 | |
210622, 25891, 311252 | |
Peptide | |
Primary | |
-20°C | |
PAMR1 |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DKFZP586H2123, FP938, Inactive serine protease PAMR1, PAMR1, RAMP, UNQ699/PRO1344 | |
Pamr1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title