Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
pan Actin Antibody (AC-40), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB12011003
Description
pan Actin Monoclonal specifically detects pan Actin in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Chicken, Chinese Hamster, Guinea Pig, Goat, Primate, Rabbit, Reptile, Sheep samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
pan Actin | |
Monoclonal | |
Unconjugated | |
Ascites with 15mM Sodium Azide | |
ACTA1 | |
Synthetic peptide: SGPSIVHALCP (human) attached to Multiple Antigen Peptide (MAP) backbone. | |
Unpurified | |
RUO | |
Primary | |
The does not react with the epitope on the N-terminal end of actin. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
AC-40 | |
Western Blot 1:500, Simple Western, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Frozen 1:10-1:500 | |
ACTA, actin, alpha 1, skeletal muscle, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin-1, ASMA, CFTD, CFTDM, MPFD, NEM1, NEM2, NEM3 | |
Mouse | |
42 kDa | |
0.1 mL | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers | |
58 | |
Human, Mouse, Rat, Pig, Amphibian, Bovine, Canine, Chicken, Chinese Hamster, Guinea Pig, Goat, Primate, Rabbit | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction