Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pan-Actin, Mouse anti-Mouse, Rat, Drosophila, Hamster, Human, Monkey, Yeast, Clone: 2A3, Millipore Sigma™

Catalog No. MABT1333100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT1333100 100 μL
MABT133325 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABT1333100 Supplier MilliporeSigma Supplier No. MABT1333100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-pan-Actin, clone 2A3, Cat. No. MABT1333, is a mouse monoclonal antibody that detects all forms of actin and has been tested for use in Western Blotting.

Actin is a multi-functional cytoskeletal protein that serves as the basic building block of cytoskeletal microfilaments. Six different actin types have been reported. Skeletal muscle alpha-actin (UniProt: P68133) is encoded by the ACTA1 gene, smooth muscle alpha-actin (UniProt: P62736) by the ACTA2 gene, cytoplasmic beta-actin (UniProt: P60709) by the ACTB gene, cardiac muscle alpha-actin (UniProt: P68032) by the ACTC1 gene, cytoplasmic gamma-actin (UniProt: P63261) by the ACTG1 gene, and smooth muscle gamma-actin (UniProt: P63267) by the ACTG2 (also known as ACTA3) gene. Although actins show about 90% overall sequence homology, isoforms do show spatial, temporal, and tissue-specific expression patterns and only 50-60% homology is found in their 18 N-terminal residues. Cytoplasmic beta and gamma actins are thought to be present in all cells, while the other four actin isoforms are typically found in specific adult muscle tissue types. Actins exist in a variety of structural states, depending on the specific ionic conditions or the interaction with ligand proteins. The oligomeric and polymeric forms that actin molecules assume are dependent on the distinct conformations they adopt. This pan-actin antibody detects all isoforms actin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen pan-Actin
Applications Western Blot
Classification Monoclonal
Clone 2A3
Conjugate Unconjugated
Dilution Western Blotting Analysis: A 1:50 dilution from a representative lot detected Actins in various species samples (e.g. Human, rat, mouse, monkey, Hamster, Drosophila, Yeast) (Courtesy of Martina Mitterhuber, M.sc and Egon Ogris, M.D., Medical University of Vienna, Austria).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Skeletal muscle alpha-actin;Smooth muscle alpha-actin;Cytoplasmic beta-actin;Cardiac muscle alpha-actin;Cytoplasmic gamma-actin;Smooth muscle gamma-actin
Gene Symbols ACTA1;ACTA2;ACTB;ACTC1;ACTG1;ACTG2
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to a fragment from the C-terminal region of human Actin.
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001092
Test Specificity Clone 2A3 detects all forms of actin in multiple species. It targets an epitope with in the C-terminal region.
Target Species Mouse, Rat, Drosophila, Hamster, Human, Monkey, Yeast
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.