Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pan Ankyrin Monoclonal Antibody (S388A-60)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527722

Catalog No. PIMA527722


Description

Description

Pan Ankyrin Monoclonal Antibody for Western Blot, ICC/IF, IHC (PFA)

Ankyrins are a family of adaptor proteins that mediate the attachment of integral membrane proteins to the spectrin-actin based membrane skeleton. Ankyrins have binding sites for the beta subunit of spectrin and at least 12 families of integral membrane proteins. This linkage is required to maintain the integrity of the plasma membranes and to anchor specific ion channels, ion exchangers and ion transporters in the plasma membrane.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pan Ankyrin
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
G5E8K5, O70511, P16157, Q01484, Q02357, Q12955, Q8C8R3
Ank1, Ank2, Ank3
Fusion protein amino acids 1-1881 (full-length) of human Ankyrin-R.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
S388A-60
Unconjugated
Ank1
2900054D09Rik; AI314020; AI835472; ANK; ANK1; Ank-1; ANK2; Ank-2; Ank3; ANK-3; AnkB; AnkG; ankyrin 1; ankyrin 1, erythrocytic; ankyrin 1, erythroid; ankyrin 2, brain; ankyrin 2, neuronal; ankyrin 3 (G); ankyrin 3, epithelial; ankyrin 3, node of Ranvier (ankyrin G); ankyrin B; ankyrin, brain; ankyrin-1; Ankyrin-2; ankyrin-2, nonerythrocytic; ankyrin-3; Ankyrin-B; ankyrin-G; Ankyrin-R; AW491075; Brain ankyrin; brain-specific ankyrin-G; brank-2; Erythrocyte ankyrin; LQT4; MRT37; nb; non-erythroid ankyrin; normoblastic anemia; pale; rCG_43073; SPH1; SPH2
Mouse
Protein G
RUO
109676, 11733, 11735, 286, 287, 288, 306570, 361833, 362036
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.