Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Pan B Cells Monoclonal Antibody (CVS36), FITC
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA528499
Description
This antibody recognizes equine Ig light chains. Characterization studies demonstrated that clone CVS36 bound 100% of CD5-ve peripheral blood lymphocytes and recognized all equine B-cells.
Pan B Cells antibody specific anti equine reagents have yet to be fully characterized for the typically recognized B-cell makers such as CD19, CD20, CD21, CD22 and CD79. While testing has demonstrated the cross reactivity of some monoclonal B-cell markers raised against other species with equine B cells, such as the Mouse anti Human CD79a antibody, (clone HM57), an overview of which may be found in the report of the second equine leucocyte antigen workshop. Pan B Cells antibody is directed against equine Ig light chains. Characterization studies demonstrated that clone CVS36 bound 100% of CD5-ve peripheral blood lymphocytes and recognized all equine B-cells (Lunn et al. 1998) As the antigen recognized by clone CVS36 appears to be present on the surface of all equine B-cells it is therefore is a reagent that can be used as a pan B-cell marker for domestic horses.
Specifications
Pan B Cells | |
Monoclonal | |
100 μg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
Purified Equine Ig. | |
100 μg | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
ELISA, Flow Cytometry | |
CVS36 | |
FITC | |
Mouse | |
Protein G | |
RUO | |
Horse | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction