Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pan Cadherin Antibody, Novus Biologicals™
SDP

Catalog No. p-5083121 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200592SS 0.025 mL
NB200592 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB200592SS Supplier Novus Biologicals Supplier No. NB200592SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 8 publications

pan Cadherin Polyclonal specifically detects pan Cadherin in Human, Mouse, Rat, Amphibian, Avian, Bovine, Canine, Chicken, Fish, Rabbit, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen pan Cadherin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:1000, Immunohistochemistry-Frozen 1:10-1:500
Formulation Whole antisera with 15mM Sodium Azide
Gene Alias Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin
Gene Symbols CDH1
Host Species Rabbit
Immunogen Synthetic peptide: DYDYLNDWGPRFKKLADMYGGGDD conjugated to KLH by a Glutaraldehyde linker. Sequence corrresponds to the C-terminal amino acids of N-Cadherin (Chicken), with an extra N-terminal lysine residue (24 amino acids).
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 999
Test Specificity Recognizes all cadherin members. Rabbit Anti-Pan Cadherin shows specific reactivity with a distinct 135 kDa band on chicken or rabbit heart extract blots using indirect immunoblotting.
Target Species Human, Mouse, Rat, Amphibian, Avian, Bovine, Canine, Chicken, Fish, Rabbit, Xenopus species
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.