Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pan-cadherin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116886
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116886 Supplier Invitrogen™ Supplier No. PA5116886
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: PC-3, A-431, Mouse brain, Rat brain Immunogen sequence: RPANPDEIGN FIDENLKAAD TDPTAPPYDS LLVFDYEGSG SEAASLSSLN SSESDKDQDY DYLNEWGNRF KKLADMYGGG EDD.

Pan Cadherin including CDH1, CDH2, CDH3, CDH4 protein belong to a family of transmembrane molecules that mediate calcium-dependent intercellular adhesion. Cadherins are involved in controlling morphogenetic movements during development and regulate cell surface adhesion through homotypic adhesion with the same cadherin species. N-cadherin's function is dependent on its association with the actin-cytoskeleton and is mediated through interactions between the C-terminal region of N-cadherin and the cytoplasmic catenin proteins. The stability of this association is regulated by phosphorylation and dephosphorylation of beta-catenin.This gene is a classical cadherin from the cadherin superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. The protein functions during gastrulation and is required for establishment of left-right asymmetry. At certain central nervous system synapses, presynaptic to postsynaptic adhesion is mediated at least in part by this gene product.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pan-cadherin
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CDH1
Gene Accession No. F1PAA9, P09803, P10287, P12830, P15116, P19022, P22223, P39038, P55283, Q63149, Q9R0T4, Q9Z1Y3
Gene Alias 2600010P09Rik; 4930525N21Rik; AA960649; AF020312; AI385538; AI851787; Arc-1; ATP-dependent helicase CHD1; ATP-dependent helicase CHD3; AW120700; AW555109; B-cadherin; CAD4; cadherin 1; cadherin 1, E-cadherin (epithelial); cadherin 1, type 1; cadherin 1, type 1 S homeolog; cadherin 1, type 1, E-cadherin (epithelial); cadherin 15, M-cadherin (myotubule); cadherin 2; cadherin 2 type 1 N-cadherin (neuronal); cadherin 2, type 1, N-cadherin (neuronal); cadherin 3; cadherin 3, P-cadherin (placental); cadherin 3, type 1, P-cadherin (placental); cadherin 4; cadherin 4 type 1 R-cadherin (retinal); cadherin 4, type 1, preproprotein; cadherin 4, type 1, R-cadherin (retinal); cadherin e; Cadherin-1; Cadherin-2; cadherin-3; Cadherin-4; cadherin-E; Cadherin-N; Cadp; calcium-dependent adhesion protein, epithelial; calcium-dependent adhesion protein, neuronal; calcium-dependent adhesion protein, placental; CAM 120/80; CD324; CD325; cdh1; cdh1.S; Cdh2; Cdh3; Cdh4; Cdh4l; CDHE; CDHN; Cdhp; CDw325; CDw325 antigen; cell-CAM 120/80; CHD1; CHD-1; CHD3; CHD-3; Chd7; chromodomain helicase DNA binding protein 1; chromodomain helicase DNA binding protein 3; chromodomain-helicase-DNA-binding protein 1; Chromodomain-helicase-DNA-binding protein 3; ECAD; E-cad; E-Cad/CTF1; E-Cad/CTF2; E-Cad/CTF3; E-cadherin; E-cadherin 1; E-cadherin precursor; Epithelial cadherin; HJMD; hZFH; K-CAM; LCAM; L-CAM; Liver cell adhesion molecule; liver cell adhesion protein; liver cell adhesion protein precursor; LOC108702627; Maternally-expressed neural cadherin; Mi-2 alpha; mi-2 autoantigen 240 kDa protein; Mi-2a; Mi2-ALPHA; Ncad; N-cadherin; N-cadherin 1; Neural cadherin; neural calcium-dependent cell adhesion molecule; Neural-cadherin; Pan Cadherin; PCAD; P-cadherin; P-cadherin (AA 232-723); Placental cadherin; Placental-cadherin; Prp7; Prp9-1; RCAD; R-CAD; R-Cadh; R-cadherin; retina cadherin; retinal cadherin; RPE-specific cadherin; Um; Unknown (protein for IMAGE:8237502); unnamed protein product; UVO; Uvomorulin; XELAEV_18024918mg; XELAEV_18046239mg; XmN-cadherin; xTCAD-1; ZFH; zinc finger helicase; zinc-finger helicase (Snf2-like)
Gene Symbols CDH1, CDH2, CDH3, CDH4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 800-882 of human pan-cadherin (NP_0043511).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1000, 1001, 1002, 100685099, 114588, 116777, 12550, 12558, 12560, 12561, 442858, 480169, 611010, 83501, 83502, 999
Target Species Human, Mouse, Rat, Canine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.