Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pan Coronavirus Spike Antibody (32), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314659AF488
This item is not returnable.
View return policy
Description
Pan Coronavirus Spike Monoclonal antibody specifically detects Pan Coronavirus Spike in Virus samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Pan Coronavirus Spike | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
This product is a recombinant monoclonal antibody expressed from HEK293 cells. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
32 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction