Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pan Coronavirus Spike Antibody (39), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314661MFV450
Description
Pan Coronavirus Spike Monoclonal antibody specifically detects Pan Coronavirus Spike in Virus samples. It is validated for Western Blot, ELISASpecifications
| Pan Coronavirus Spike | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Coronavirus Spike S1+S2 ECD protein. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| 39 | |
| Western Blot, ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction