Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pan Cytokeratin Mouse anti-Human, Clone: PAN-CK (Cocktail), Abnova™

Catalog No. 89034745 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-745 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-745 Supplier Abnova Corporation Supplier No. MAB14662
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against native human Pan Cytokeratin.

  • High specificity
  • High reactivity

Specifications

Antigen Pan Cytokeratin
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone PAN-CK (Cocktail)
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native human Pan Cytokeratin.
Dilution Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by
Formulation In PBS.
Gene Accession No. P08729; Q04695; P05783; Q7Z794; Q01546
Gene Alias CK7/K2C7/K7/MGC129731/MGC3625/SCL
Gene Symbols KRT7
Host Species Mouse
Immunogen Human epidermal keratin.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3855/3872/3875/374454/51350
Target Species Human
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.