Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Pan DIDO, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN1367MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1367MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1367MI Supplier MilliporeSigma™ Supplier No. ABN1367
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Death-inducer obliterator 1 (UniProt Q9BTC0; also known as DATF-1, Death-associated transcription factor 1, DIO-1, hDido1) is encoded by the DIDO1 (also known as C20orf158, DATF1, KIAA0333) gene (Gene ID 11083) in human. The DIDO1 gene encodes four alternatively spliced variants (DIDO1/isoform 2, DIDO2/isoform 1, DIDO3/isoform 4, isoform a/isoform 3) implicated to be involved in apoptosis and development. The longest and most widely expressed DIDO3 is assigned as the canonical isoform. DIDO3 is a nuclear protein that forms part of the spindle assembly checkpoint (SAC) and is necessary for correct chromosome segregation in somatic and germ cells. Dido3-knockout compromises differentiation of murine embryonic stem cells (mESCs) in vitro and of epiblast cells in vivo, resulting in early embryonic death at around day 8.5 of gestation. On the other hand, exogenous expression of Dido1 isoform in mESCs is reported to inhibit LIF withdrawal-induced differentiation, while Dido1 knockdown facilitate mESCs differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pan DIDO
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene DIDO1, C20orf158, DATF1, KIAA0333
Gene Accession No. Q9BTC0
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding an N-terminal sequence of human DIDO1.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_071388
Test Specificity This polyclonal antibody detects an N-terminal epitope present in all four spliced isoforms of human DIDO1 reported by UniProt (Q9BTC0).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.