Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pan Endothelial Cell Marker Antibody (RM0061-4J13), Alexa Fluor™ 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212438AF488
Description
Pan Endothelial Cell Marker Monoclonal specifically detects Pan Endothelial Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Pan Endothelial Cell Marker | |
Monoclonal | |
Alexa Fluor 488 | |
Rat | |
Protein A or G purified | |
RUO | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0061-4J13 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Cell lysates of mouse endothelial cells | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction