Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pan Endothelial Cell Marker Antibody (RM0061-4J13) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $560.50
Specifications
| Antigen | Pan Endothelial Cell Marker |
|---|---|
| Clone | RM0061-4J13 |
| Concentration | LYOPH |
| Applications | Immunohistochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), CyTOF, Immunoprecipitation |
| Classification | Monoclonal |
Description
Pan Endothelial Cell Marker Monoclonal specifically detects Pan Endothelial Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Pan Endothelial Cell Marker | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Cell lysates of mouse endothelial cells | |
| Primary |
| RM0061-4J13 | |
| Immunohistochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), CyTOF, Immunoprecipitation | |
| Unconjugated | |
| Rat | |
| Mouse | |
| IgG2 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title