Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pan-Endothelium Marker, p Ser123, p Ser124 Antibody (HIS52), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200076H
Description
Pan-Endothelium Marker Monoclonal antibody specifically detects Pan-Endothelium Marker in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot (Negative)Specifications
Pan-Endothelium Marker | |
Monoclonal | |
HRP | |
PBS | |
The immunogen for this antibody is Pan endothelium. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
HIS52 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Western Blot (Negative) | |
Mouse | |
Protein A or G purified | |
RUO | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction