Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAN Indicator, 0.1% Solution in Ethanol, Spectrum™ Chemical
SDP

Catalog No. p-7158608 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle

Catalog No. 18-604-955

Supplier: Spectrum Chemical Manufacturing Corporation P575100ML

Only null left
Add to Cart
Add to Cart

P-575, 85-85-8

Spectrum™ Chemical PAN Indicator, 0.1 Percent Solution in Ethanol

Chemical Identifiers

CAS 85-85-8, 67-56-1
Molecular Formula C15H11N3O
Molecular Weight (g/mol) 249.27
InChI Key RAXUMGMWXZYADR-SDXDJHTJSA-N
IUPAC Name (1Z)-1-[2-(pyridin-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
SMILES O=C1C=CC2=CC=CC=C2\C1=N\NC1=CC=CC=N1

Specifications

CAS Max % 0.999, 0.001
Packaging Poly Bottle
Quantity 100 mL
UN Number UN1170
Grade Chemical Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.