Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAN Indicator, 0.1% w/v in Methanol, Reagents
Click to view available options
Quantity:
500 mL
Specifications
Specifications
Chemical Name or Material | PAN Indicator, 0.1% w/v in Methanol |
CAS | 67-56-1, 85-85-8 |
Boiling Point | 64.5°C |
Synonym | 1-2-Pyridylazo-2-naphthol, 1-(Pyridin-2-yldiazenyl)naphthalen-2-ol |
Percent Purity | 0.1% |
Grade | Reagent |
Packaging | Glass Bottle |
Specific Gravity | 0.7958 |
Color | Orange/Yellow |
Quantity | 500 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction