Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAN Indicator, 0.3% in Ethanol, Certified, LabChem™

Catalog No. p-5389132
Click to view available options
Quantity:
125 mL
500 mL

Specifications

Chemical Name or Material PAN Indicator
Name Note 0.3% in Ethanol
CAS Max % 89.7, 5, 5, 0.3
CAS 67-56-1, 67-63-0, 85-85-8
Linear Formula C15H11N3O
UN Number UN1170
Solubility Information Soluble in water
Identification Passes Test
Formula Weight 249.27
Grade Certified
Packaging Amber Glass
Suitability Passes Test
Decomposition Information Carbon monoxide; Carbon dioxide; May release flammable gases
Color Orange to Red
Quantity 125 mL
Physical Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.