Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAN Indicator, 0.3% (w/v) in Reagent Alcohol, Ricca Chemical

Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
Specifications
Specifications
Chemical Name or Material | PAN Indicator |
CAS | 64-17-5, 67-63-0, 67-56-1, 85-85-8 |
CAS Min % | 88.0, 4.0, 4.0, 0.37 |
CAS Max % | 92.0, 6.0, 6.0, 0.38 |
Percent Purity | 0.3% W/V |
Grade | Indicator |
Concentration or Composition (by Analyte or Components) | 0.3% (w/v) in Ethanol |
Packaging | Amber Glass Bottle |
Color | Amber |
Quantity | 120 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction