Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 1 Antibody Cocktail
GREENER_CHOICE

Catalog No. PIMA549309
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549309 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549309 Supplier Invitrogen™ Supplier No. MA549309
Add to Cart
Add to Cart

Antibody Cocktail

Adding 1 mL of PBS buffer will yield a concentration of 100 μg/mL. Positive Control - WB: human NCL-H460 whole cellhuman Hela whole cellhuman A431 whole cell. IHC: Human Placenta tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between approximately40 kDa and approximately68 kDa and isoelectric pH between 4. 9 - 7. 8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Cocktail
Clone C-11+PCK-26+CY-90+KS-1A3+M20+A53-B/A2
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation 1.2% sodium acetate with 2mg BSA and 0.01mg sodium azide
Gene KRT1
Gene Accession No. P04264, Q6IMF3
Gene Alias 67 kDa cytokeratin; CK1; CK-1; cytokeratin 1; cytokeratin-1; EHK; EHK1; epidermolytic hyperkeratosis 1; epithelial keratin 1; Epithelial keratin-1; EPPK; Hair alpha protein; K1; Kb1; KER1; keratin 1; keratin 1 (epidermolytic hyperkeratosis); keratin 1, type II; keratin 86; keratin complex 2, basic, gene 1; keratin, type II cytoskeletal 1; Keratin-1; KRT1; krt1 {ECO:0000250; KRT1A; Krt-2.1; Krt2-1; Krt86; KRTA; NEPPK; type II keratin Kb1; type-II keratin Kb1; UniProtKB:P04264}
Gene Symbols KRT1
Host Species Mouse
Immunogen mixture of several monoclonal cytokeratin clones.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300250, 3848
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG1, IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.