Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pan methylated lysine Mouse, Clone: , Abnova™
Mouse monoclonal antibody raised against pan methylated lysine.
Supplier: Abnova Corporation MAB12422
Specifications
Pan methylated lysine | |
Monoclonal | |
Mouse monoclonal antibody raised against pan methylated lysine. | |
In PBS, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
Methylated BSA | |
100 μL | |
Primary | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:200-1:500) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user. | |
Mouse | |
Affinity chromatography | |
RUO | |
Pan methylated lysine monoclonal antibody detects endogenous pan methylated lysine protein. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction