Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pan Nuclear Pore Complex Marker Antibody (39C7), Novus Biologicals™
SDP

Catalog No. NBP105390 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105390 0.5 mL
NB018368 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP105390 Supplier Novus Biologicals Supplier No. NBP105390
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

pan Nuclear Pore Complex Marker Monoclonal specifically detects pan Nuclear Pore Complex Marker in Human, Mouse, Rat, Porcine, Bovine, C. elegans, Chicken, Drosophila, Equine, Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen pan Nuclear Pore Complex Marker
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 39C7
Conjugate Unconjugated
Dilution Western Blot 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:500
Formulation Supplied as concentrated hybridoma cell culture media. with 5mM Sodium Azide
Gene Alias 107 kDa nucleoporin, NPC proteins, Nuclear pore complex protein Nup107, Nucleoporin 107kDa, Nucleoporin Nup107, NUP 107, NUP 84, NUP107, NUP84
Host Species Mouse
Immunogen Yeast nuclear preparations
Molecular Weight of Antigen 62 kDa
Purification Method Tissue culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Test Specificity Panspecific
Target Species Human, Mouse, Rat, Pig, Bovine, C. elegans, Chicken, Drosophila, Equine, Yeast
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.