Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pan-PMCA Mouse anti-Chicken, Rat, Human, Unconjugated, Clone: 5F10, MilliporeSigma™

Catalog No. MABN180225 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN1802-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1802-25 Supplier MilliporeSigma Supplier No. MABN180225UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-pan-PMCA, clone 5F10, Cat. No. MABN1802, is a mouse monoclonal antibody that targets pan-Plasma membrane calcium-transporting ATPase (PMCA) and has been tested in ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen pan-PMCA
Applications ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5F10
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Plasma membrane calcium-transporting ATPase;Plasma membrane calcium pump
Gene Symbols ATP2B1;ATP2B2;ATP2B3;ATP2B4
Host Species Mouse
Immunogen Purified human erythrocyte calcium ATPase.
Molecular Weight of Antigen 134 to 139 kDa calculated.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 5F10 detects all isoforms of Plasma membrane calcium-transporting ATPase (PMCA) in human, mouse, and rat.
Target Species Chicken, Rat, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.