Learn More
Invitrogen™ Pan Ras Polyclonal Antibody

Description
Positive Control: A549, H1299, HCT116, KRAS, HRAS, NRAS, zebrafish liver, zebrafish GI, whole zebrafish Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Pan Ras |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.27 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | Nras |
| Gene Accession No. | P01111, P01112, P01116, P08556, P08644, P20171, P32883, P79737, Q04970, Q61411 |
| Gene Alias | AI929937; ALPS4; AV095280; C-BAS/HAS; cellular c-Ki-ras2; cellular c-Ki-ras2 proto-oncogene; CFC2; c-Ha-ras; c-Ha-ras p21 protein; c-Ha-ras transgene; C-HA-RAS1; c-has/bas p21 protein; C-H-RAS; c-Ki-ras; c-Kirsten-ras protein; c-Kirsten-ras proto-oncogene; c-K-ras; c-K-ras2 protein; CMNS; c-rasHa; c-ras-Ki-2 activated oncogene; CTLO; etID18709.8; fa04e08; fc14b12; fc23g10; fj89d12; fj99a10; GTP- and GDP-binding peptide B; GTPase HRas; GTPase HRas, N-terminally processed; GTPase Hras1; GTPase KRas; GTPase KRas, N-terminally processed; GTPase NRas; HAMSV; Ha-ras; Ha-Ras1 proto-oncoprotein; Harvey ras1 protein; Harvey rat sarcoma viral (v-Ha-ras) oncogene homolog; Harvey rat sarcoma viral oncogene homolog; Harvey rat sarcoma viral oncoprotein; Harvey rat sarcoma virus oncogene; Harvey rat sarcoma virus oncogene 1; Harvey-ras; hras; H-ras; H-ras 1 protein; HRas proto-oncogene, GTPase; HRas proto-oncogene, GTPase a; HRAS1; H-Ras-1; Hras-1; hrasa; H-RASIDX; hypothetical protein LOC445289; hypothetical protein LOC550286; K RAS; Ki-ras; Kirsten rat sarcoma oncogene 2, expressed; Kirsten rat sarcoma viral oncogene; Kirsten rat sarcoma viral oncogene homolog; Kirsten rat sarcoma viral oncogene homologue 2 (active); Kras; K-ras; K-Ras 2; K-ras p21 protein; KRAS proto-oncogene, GTPase; KRAS1; KRAS2; Kras-2; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; NCMS; Neuroblastoma RAS; neuroblastoma ras oncogene; neuroblastoma RAS viral (v-ras) oncogene homolog; neuroblastoma RAS viral oncogene homolog; NRAS; N-ras; N-ras oncogene p21; N-ras protein part 4; NRAS proto-oncogene, GTPase; NRAS1; NS; NS3; NS6; oncogene KRAS2; p19 H-RasIDX protein; p21; p21 N-ras oncogene; p21 protein; p21B; p21ras; PR310 c-K-ras oncogene; RALD; ras; Ras family small GTP binding protein H-Ras; RASH1; RASK; RASK2; ras-like protein; RASN; sb:eu826; transformation gene: oncogene HAMSV; transforming protein N-Ras; Transforming protein p21; v-Ha-ras Harvey rat sarcoma viral oncogene homolog; v-Ha-ras Harvey rat sarcoma viral oncogene homolog a; v-Ki-ras2 Kirsten rat sarcoma 2 viral oncogene homolog; v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog; v-ras neuroblastoma RAS viral oncogene homolog; wu:fa04e08; wu:fc14b12; wu:fc23g10; wu:fj89d12; wu:fj99a10; xRAS1; xRAS2; zgc:100761; zgc:110250; zgc:85725; zras-b1 |
| Gene Symbols | HRAS, hrasa, KRAS, Nras |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.