Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pan SHANK Monoclonal Antibody (N23b/49), APC
GREENER_CHOICE

Catalog No. PIMA545438
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545438 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545438 Supplier Invitrogen™ Supplier No. MA545438
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45438 was sufficient for detection of Shank1-4 in 10 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 160kDa. Recognizes Shank1, 2 and 3.

Shank proteins make up a family of scaffold proteins identified through their interaction with a variety of membrane and cytoplasmic proteins. Shank proteins at postsynaptic sites of excitatory synapses play roles in signal transmission into the postsynaptic neuron. Studies suggest that Shank2 is expressed in the neurons of the developing retina, and could play a role in the neuronal differentiation of the developing retina. Other recent studies suggest that the disruption of glutamate receptors at the Shank postsynaptic platform could contribute to the destruction of the postsynaptic density, which underlies the synaptic dysfunction and loss in Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pan SHANK
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry, Microarray
Classification Monoclonal
Clone N23b/49
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene SHANK2
Gene Accession No. D3YZU1, Q4ACU6, Q80Z38, Q9BYB0, Q9JLU4, Q9QX74, Q9UPX8, Q9WV48, Q9Y566
Gene Alias AI841104; AUTS17; cortactin SH3 domain-binding protein; cortactin-binding protein 1; CORTBP1; CTTNBP1; DEL22q13.3; GKAP/SAPAP interacting protein; GKAP/SAPAP-interacting protein; KIAA1022; Kiaa1650; mKIAA1022; proline rich synapse associated protein 1; proline rich synapse associated protein 2; proline-rich synapse associated protein 1; proline-rich synapse-associated protein 1; Proline-rich synapse-associated protein 2; ProSAP1; Prosap2; PSAP2; SAPAP-interacting protein synamon; SCZD15; SH3 and multiple ankyrin repeat domains 1; SH3 and multiple ankyrin repeat domains 2; SH3 and multiple ankyrin repeat domains 3; SH3 and multiple ankyrin repeat domains protein 1; SH3 and multiple ankyrin repeat domains protein 2; SH3 and multiple ankyrin repeat domains protein 3; SH3 and multiple ankyrin repeat domains protein 3 variant c-3; SH3 and multiple ankyrin repeat domains protein 3 variant c-4; SH3 and multiple ankyrin repeat domains-like protein 3 variant 1; SH3 and multiple ankyrin repeat domains-like protein 3 variant 2; SH3 and multiple ankyrin repeat domains-like protein 3 variant 3; SH3 and multiple ankyrin repeat domains-like protein 3 variant c-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant c-2; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-2; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-3; SH3 and multiple ankyrin repeat domains-like protein 3 variant e-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant e-2; SH3/ankyrin domain gene 1; SH3/ankyrin domain gene 2; SH3/ankyrin domain gene 3; SHANK; shank postsynaptic density protein; Shank postsynaptic density protein 3a; SHANK1; Shank1a; Shank2; Shank3; shank3 splice variant II; shank3 splice variant III; shank3 splice variant IV; Shank3b; SHANK3c-3; SHANK3c-4; shank3e splice variant III; shank3e splice variant IV; somatostatin receptor-interacting protein; Spank1; SPANK-1; SPANK-2; SPANK-3; SSTR-interacting protein; Sstrip; synamon
Gene Symbols Shank1, SHANK2, Shank3
Host Species Mouse
Immunogen Fusion protein amino acids 84-309 of rat Shank2.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171093, 210274, 22941, 243961, 50944, 58234, 59312, 78957, 85358
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.