Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Pan Shc, Mouse, Unlabeled, Clone: 3A12.1,

Catalog No. MABN1596MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1596MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1596MI Supplier MilliporeSigma™ Supplier No. MABN1596
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SHC-transforming protein 1 (UniProt P29353; also known as SH2 domain protein C1, SHC-transforming protein 3, SHC-transforming protein A, Src homology 2 domain-containing-transforming protein C1) is encoded by the SHC1 (also known as SHC, SHCA) gene (Gene ID 6464) in human. Originally identified as an SH2-containing proto-oncogene involved in growth factor signaling, Shc proteins function as adaptor molecules recruited by numerous receptors (e.g. integrins, GPCRs, and receptors for growth factors, antigens, cytokines, and hormones) for signling. In addition to the ubiquitously expressed Shc (a.k.a. ShcA by SHC1/SHCA gene), there exist also ShcB (by SHC2/SHCB gene) and ShcC (by SHC3/SHCC gene), which are predominantly expressed in neuronal cells. Alternative splicings result in six ShcA isoforms, including p66Shc, p52Shc, and p46Shc. All ShcA isoforms possess two distinct domains, PTB (a.a. 156-339) and SH2 (a.a. 488-579), that bind phosphotyrosine-containing sequences, as well as a central region (CH1; a.a. 340-487) that contains critical tyrosine phosphorylation sites. The phosphotyrosine-binding (PTB) domain is also referred to as phosphotyrosine-interaction domain or PID.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pan Shc
Applications Western Blot
Classification Monoclonal
Clone 3A12.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene SHC1, SHC, SHCA
Gene Accession No. P29353
Host Species Mouse
Immunogen GST-tagged recombinant human Shc C-terminal region fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001123512
Test Specificity Clone 3A12.1 was raised against a C-terminal region sequence present in all six human SHC1 spliced isoforms reported by UniProt (P29353).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.