Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ pan Synuclein Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578420
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578420 Supplier Invitrogen™ Supplier No. PA578420
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: mouse brain, rat brain Predicted Reactivity: Human (93%), Xenopus laevis (84%), Pig (92%), Chicken (87%), Sheep (91%), Chimpanzee (93%), Bovine (91%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Synucleins are small, soluble proteins expressed primarily in neural tissue and in certain tumors. The family includes three known proteins: alpha-synuclein, beta-synuclein, and gamma-synuclein. The alpha- and beta-synuclein proteins are found primarily in brain tissue, where they are seen mainly in presynaptic terminals. The gamma-synuclein protein is found primarily in the peripheral nervous system and retina, but its expression in breast tumors makes it a candidate as a marker for tumor progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen pan Synuclein
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene SNCA
Gene Accession No. O55042, P37377, Q63544, Q63754, Q91ZZ3, Q9Z0F7
Gene Alias AI838531; alphaSYN; alpha-synuclein; BCSG1; betaSYN; Beta-synuclein; Breast cancer-specific gene 1 protein; C79089; gamma-synuclein; NACP; non A-beta component of AD amyloid; non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; PARK1; PARK4; PD1; persyn; phosphoneuroprotein 14; PNP 14; PRSN; Sensory neuron synuclein; Snca; SNCB; SNCG; SR; Syn; Synoretin; synuclein alpha; synuclein alpha-140; synuclein beta; synuclein gamma; synuclein, alpha; synuclein, alpha (non A4 component of amyloid precursor); synuclein, beta; synuclein, gamma; synuclein, gamma (breast cancer-specific protein 1)
Gene Symbols SNCA, SNCB, SNCG
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of Rat pan Synuclein. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104069, 113893, 20617, 20618, 29219, 64347
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.