Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ pan T cells Monoclonal Antibody (OX-52), Biotin

Catalog No. PIMA517532
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517532 500 μg
PIMA517531 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA517532 Supplier Invitrogen™ Supplier No. MA517532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-rat pan T lymphocyte monoclonal antibody immunoprecipitates a two chain structure (95, 120 kDa) largely restricted to T lymphocytes and thymocytes. Applications therefore include the identification of T lymphocyte lineage cells in suspension and tissue. The antibody has previously been used to stain T cell areas of the spleen, lymph nodes and Peyer's patch. In the thymus, this antibody labels all thymocytes, however medullary cells are more strongly positive than cortical cells. The antibody does not label such non-lymphoid tissues such as brain, kidney, liver and skin. This antibody stains approximately 1.0% of bone marrow cells, and 56% thoracic duct lymphocytes. Weak staining occurs with 50% dendritic cells from thoracic duct of mesenteric lymphadenectomized rats. The antigen recognized by this antibody is not expressed on granulocytes or macrophages. The function of the antigen recognized by this antibody has not, as yet, been associated with any particular function of T cells. The antibody does not inhibit the allogeneic mixed leukocyte response, nor does it inhibit T cytotoxic effector cell function. This clone works in flow cytometry, Immunoprecipitation and with frozen sections.

Pan T cells, encompassing all T lymphocyte subsets, are integral to the adaptive immune system, originating from hematopoietic stem cells in the bone marrow and maturing in the thymus. These cells include helper T cells (CD4+), cytotoxic T cells (CD8+), regulatory T cells, and memory T cells, each playing distinct roles in immune defense. Pan T cells are characterized by the expression of the T-cell receptor (TCR) complex, which enables antigen recognition and response. The diversity within pan T cells allows for a broad range of functions, from direct cytotoxic effects on infected or malignant cells to regulatory roles in maintaining immune tolerance and memory responses for long-term immunity. These cells also interact with other immune components, enhancing or modulating responses to pathogens and tumors. Pan T cells are pivotal in cancer immunotherapy, where targeting specific T cell subsets can enhance anti-tumor immunity and overcome resistance mechanisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen pan T cells
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone OX-52
Concentration 0.1 mg/mL
Conjugate Biotin
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Host Species Mouse
Immunogen Immunogen:PVG-RT1c thoracic duct lymphocytes and spleen cells.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.