Learn More
Invitrogen™ Pan TEF1 Recombinant Rabbit Monoclonal Antibody (3Q10Q8)

Description
Immunogen sequence: SHDVKPFSQQ TYAVQPPLPL PGFESPAGPA PSPSAPPAPP WQGRSVASSK LWMLEFSAFL EQQQDPDTYN KHLFVHIGQS SPSYSDPYLE.
Specifications
Specifications
| Antigen | Pan TEF1 |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 3Q10Q8 |
| Concentration | 2.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Tead1 |
| Gene Accession No. | P28347 |
| Gene Alias | 2610024B07Rik; AA; B230114H05Rik; Gtrgeo5; mTEF-1; N TEAD1; NTEF1; NTEF-1; protein GT-IIC; REF1; Tcf13; TCF-13; TEA domain family member 1; TEA domain family member 1 (SV40 transcriptional enhancer factor); TEA domain transcription factor 1; TEAD1; TEAD-1; TEF1; TEF-1; Transcription factor 13; transcriptional enhancer factor 1; transcriptional enhancer factor TEF-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.