Learn More
Invitrogen™ PAN2 Polyclonal Antibody

Description
Recommended positive controls: Jurkat. Predicted reactivity: Mouse (99%), Rat (98%), Chicken (93%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | PAN2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Pan2 |
| Gene Accession No. | Q504Q3 |
| Gene Alias | 1200014O24Rik; AI047843; AN2 polyA specific ribonuclease subunit homolog; AW742773; HAMAP-Rule:MF_03182}; Inactive ubiquitin carboxyl-terminal hydrolase 52; inactive ubiquitin carboxyl-terminal hydrolase 52 {ECO:0000255; Kiaa0710; mKIAA0710; PAB1P-dependent poly(A)-nuclease; PAB1P-dependent poly(A)-nuclease {ECO:0000255; PAB1P-dependent poly(A)-specific ribonuclease; PAB-dependent poly(A)-specific ribonuclease subunit 2; PAB-dependent poly(A)-specific ribonuclease subunit PAN2; PAB-dependent poly(A)-specific ribonuclease subunit PAN2 {ECO:0000255; PABP-dependent poly(A) nuclease 2; PAN deadenylation complex catalytic subunit 2; PAN deadenylation complex catalytic subunit 2 {ECO:0000255; PAN deadenylation complex subunit 2; Pan2; PAN2 poly(A) specific ribonuclease subunit; PAN2 poly(A) specific ribonuclease subunit homolog; PAN2 polyA specific ribonuclease subunit; PAN2 polyA specific ribonuclease subunit homolog; PAN2 polyA specific ribonuclease subunit homolog (S. cerevisiae); PAN2-PAN3 deadenylation complex catalytic subunit Pan2; Poly(A)-nuclease deadenylation complex subunit 2; ubiquitin specific peptidase 52; ubiquitin specific protease 52; Usp52 |
| Gene Symbols | Pan2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.