Learn More
PAN2/USP52 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | PAN2/USP52 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-8932
![]() |
Bethyl Laboratories, Inc
A304882AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
This gene encodes a deadenylase that functions as the catalytic subunit of the polyadenylate binding protein dependent poly(A) nuclease complex. The encoded protein is a magnesium dependent 3' to 5' exoribonuclease that is involved in the degradation of cytoplasmic mRNAs. Alternate splicing results in multiple transcript variants.Specifications
PAN2/USP52 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
USP52, PAB-dependent poly(A)-specific ribonuclease subunit 2, PABP-dependent poly(A) nuclease 2, PAN2 polyA specific ribonuclease subunit, inactive ubiquitin carboxyl-terminal hydrolase 52, ubiquitin specific peptidase 52, ubiquitin specific protease 52 | |
Between 1 and 50 | |
Primary | |
4° C | |
PAN2 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q504Q3, Q8BGF7 | |
103135, 9924 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.