Learn More
PAN2/USP52 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304882A
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
This gene encodes a deadenylase that functions as the catalytic subunit of the polyadenylate binding protein dependent poly(A) nuclease complex. The encoded protein is a magnesium dependent 3' to 5' exoribonuclease that is involved in the degradation of cytoplasmic mRNAs. Alternate splicing results in multiple transcript variants.Specifications
| PAN2/USP52 | |
| Polyclonal | |
| Unconjugated | |
| PAN2 | |
| USP52, PAB-dependent poly(A)-specific ribonuclease subunit 2, PABP-dependent poly(A) nuclease 2, PAN2 polyA specific ribonuclease subunit, inactive ubiquitin carboxyl-terminal hydrolase 52, ubiquitin specific peptidase 52, ubiquitin specific protease 52 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 103135, 9924 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q504Q3, Q8BGF7 | |
| PAN2 | |
| Between 1 and 50 | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.